Skip to Content
Merck
All Photos(2)

Key Documents

5.33919

Sigma-Aldrich

Galactokinase Inhibitor, Cpd36

Synonym(s):

Galactokinase Inhibitor, Cpd36, 2-(Benzo[d]oxazol-2-ylamino)-4-(4-chloro-1H-pyrazol-3-yl)-4,6,7,8-tetrahydroquinazolin-5(1H)-one, GALK1 Inhibitor, Cpd36

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H15ClN6O2
Molecular Weight:
382.80
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 20 mg/mL

storage temp.

−20°C

General description

A cell-permeable tetrahydroquinazolinone derivative that acts as a highly selective inhibitor of galactokinase 1 (GALK1, IC50 = 4.7 µM in a GALK-ATP depletion assay). The inhibition appears to be competitive with respect to ATP and uncompetitive with respect to galactose. Exhibits higher affinity for GALK1-galactose complex than the free enzyme. Does not affect GALK2 activity even at higher concentrations (~ 40 µM) and has no significant effect on all other protein kinases profiled. Shown to reduce Gal-1-phosphate levels in patient derived fibroblasts without affecting their viability. Displays good plasma (t1/2 >60 min) and microsomal stability (t1/2
Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.
A cell-permeable tetrahydroquinazolinone derivative that acts as a highly selective inhibitor of galactokinase 1 (GALK1, IC50 = 4.7 µM in a GALK-ATP depletion assay). The inhibition appears to be competitive with respect to ATP and uncompetitive with respect to galactose. Exhibits higher affinity for GALK1-galactose complex than the free enzyme. Does not affect GALK2 activity even at higher concentrations (~ 40 µM) and has no significant effect on all other protein kinases profiled. Shown to reduce Gal-1-phosphate levels in patient derived fibroblasts without affecting their viability. Displays good plasma (t1/2 >60 min) and microsomal stability (t1/2


Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
GALK1
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Liu, L., et al. 2015. Bioorg. Med. Chem. Lett.25, 721.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service