Key Documents
TEM-LC300AU
Sigma-Aldrich
Lacey Carbon Supported Gold Grids
size 300 mesh, size 50 nm , hole size, box of 5 ×
Synonym(s):
Lacey Carbon Support Film, 300 mesh Au Grids, 5pk
About This Item
UNSPSC Code:
41111761
packaging
box of 5 ×
size
300 mesh
50 nm , hole size
General description
Lacey carbon film supported TEM grids. This carbon coating consists of woven-mesh-like openings of different sizes and shapes. This type of pattern provides support but does not interfere when observing specimen sections.
Choose from one of the most recent versions:
Certificates of Analysis (COA)