DWK218062405
DURAN® graduated amber laboratory bottle
bottle capacity 100mL, GL 45 thread, without screw cap and pouring ring, borosilicate glass 3.3 bottle
About This Item
material
amber
borosilicate glass 3.3 bottle
Joint
joint threaded neck
sterility
non-sterile
feature
autoclavable
graduated
without cap
without screw cap and pouring ring
packaging
pack of 10 ea
manufacturer/tradename
DWK Life Sciences DURAN® 218062405
bottle capacity
100mL
bottle diam. × H
56 mm × 100 mm
fitting
GL 45 thread
General description
Application
DURAN® graduated laboratory bottles are suitable for the storage of reagents, chemicals, solutions, tissue culture media, and more.
Features and Benefits
- Manufactured from ambered DURAN® borosilicate glass 3.3. DURAN® is a neutral glass, classified as a Glass Type I according to USP <660>, EP (3.2.1) and JP <7.01>, and meets ASTM E438, Type I, Class A requirements
- Standardised GL-thread and matching cap system for particularly tight sealing and simple, clean pouring out
- USP <660> (Spectral Transmission) and EP (3.2.1) compliant. Less than 10% spectral transmission between 290 nm and 450 nm.
- The only external application of the colour ensures that internal surfaces have standard DURAN® glass properties
- Very durable and chemically resistant amber colour
- Complete with blue screw cap (PP, integral lip seal) and pouring ring (PP) for drip-free pouring and clean, safe working
- Easy-to-read scale with graduation marks to estimate the content
- Fired-on, highly durable white ceramic print
- Sizes 10 and 25 ml with specially shaped glass edge for improved pouring out whereby an additional plastic pouring ring is not required
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)