DWK211856302
Duran® Bottles, Reagent, Wide Neck
clear bottle (reagent)
Synonym(s):
211856302
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
Recommended Products
material
clear bottle (reagent)
flat head glass cap (stopper)
wide-neck
Joint
Joints ST/NS neck (NS 60/46)
packaging
pkg of 10 ea
manufacturer/tradename
DURAN® 211856302
capacity
2000 mL
Looking for similar products? Visit Product Comparison Guide
General description
The classical stoppered reagent bottles are moulded from DURAN® borosilicate 3.3 Type 1 glass, and are suitable for the storage of liquid and solid chemicals and reagents. The bottles feature a wide conical ground joint neck and comply with ISO 4796-2. The wider neck is more convenient for handling powders. The glass stopper provides an excellent seal that protects the contents from evaporation, oxidation, and contamination. The bottles have a Retrace Code for manufacturing lot traceability and a marking field in permanent white enamel.
Application
Autoclaving of media and reagents, Preparation of media and reagents, Sample collection, Culturing vessel, Storage of media and reagents, Transport and shipping, Liquid handling reservoir, Boro 3.3
Features and Benefits
- Manufactured from borosilicate glass 3.3
- Easy access to the bottle contents due to the wide diameter of the bottle neck
- DURAN® borosilicate glass 3.3. DURAN® is a neutral glass, classified as a Glass Type I according to USP <660>, EP (3.2.1) and JP <7.01>, and meets ASTM E438, Type I, Class A requirements
- All borosilicate glass components and therefore suitable for storage of aggressive materials
- Tight closure due to standard taper ground glass joint
- Fired-on, highly durable white ceramic print
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service