Skip to Content
Merck
All Photos(1)

Key Documents

CF0026

Sigma-Aldrich

Pyrazolyltetrafluoroethyl trimethylsilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C8H12F4N2Si
CAS Number:
MDL number:
UNSPSC Code:
12352101

form

liquid

reaction suitability

reaction type: C-C Bond Formation

Application

After being activated with silicophilic activators, like fluorides, alkoxides or carboxylates, pyrazolyltetrafluoroethyl trimethylsilane serves as a nucleophilic source of the pyrazolyltetrafluoroethyl moiety in reaction with electrophiles such as aromatic aldehydes.

Automate your fluorination reactions with Synple Automated Synthesis Platform (SYNPLE-SC002)

Legal Information

Product of CF Plus Chemicals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

The fluoroalkylation toolbox now includes Togni reagents, hypervalent iodine perfluoroalkylation reagents, fluoroalkyl bromides, silanes, carboxylates, and sulfonyl fluorides for late stage fluoroalkylation.

The fluoroalkylation toolbox now includes Togni reagents, hypervalent iodine perfluoroalkylation reagents, fluoroalkyl bromides, silanes, carboxylates, and sulfonyl fluorides for late stage fluoroalkylation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service