Documentos clave
Z154792
Sigma-Aldrich
VARITEMP® heat gun
AC/DC input 120 V AC
About This Item
UNSPSC Code:
41102404
manufacturer/tradename
VT-750C
availability
not available in EU
parameter
≤1000 °F temperature (538 °C)
output
14.5 A
General description
Dial-in and lock temperature control (ambient to maximum); 3-position rocker switch (off/cold/hot); rubber-backed base with 90° rotation; heavy-duty 3-wire line cord and plug on 120 V unit (220/240 V unit supplied without plug); mica-insulated ceramic heating element; double-jacketed die-cast aluminum housing.
Legal Information
VARITEMP is a registered trademark of Master Appliance Corp.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)