Skip to Content
Merck
All Photos(1)

Key Documents

SCP0035

Sigma-Aldrich

[Gln11]-Amyloid β 1-28 human

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C145H210N42O45
Molecular Weight:
3261.47
UNSPSC Code:
12352209
NACRES:
NA.32

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥65%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Gln-Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser-Asn-Lys

Application

Amyloid β (Aβ) refers to peptides derived from Amyloid precursor protein that vary in length from 36-43 amino acids. Aβ(s) peptides, their peptide fragments and mutated fragments are used to study a wide range of metabolic and regulatory functions including activation of kinases, regulation of cholesterol transport, function as a transcription factor, and regulators of inflammation. Aβ(s) peptides and their peptide fragments are also used to study oxidative stress, metal binding and mechanisms of protein cross-linking in the context of diseases such as Alzheimer′s disease and neurodegeneration.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phosphorus Dendrimers Affect Alzheimer's (Aμ(1-28)) Peptide and MAP-Tau Protein Aggregation.
Wasiak T, Ionov M, Nieznanski K, et al.
Molecular Pharmacology, 9(3), 458-469 (2012)
Daniela Valensin et al.
Inorganic chemistry, 50(15), 6865-6867 (2011-07-02)
The reactions of human β-amyloid peptide 1-28 (Aβ28) with Al(III) and Fe(III) ions were investigated by (1)H NMR and electrospray ionization mass spectrometry (ESI-MS) under pH conditions close to physiological ones. (1)H NMR titrations, performed in the 5.3-8.0 pH range
Daniela Valensin et al.
Inorganic chemistry, 49(11), 4720-4722 (2010-05-13)
The reaction of the ruthenium(II) complex fac-[Ru(CO)(3)Cl(2)(N(1)-thz)] (I hereafter; thz = 1,3-thiazole) with human beta-amyloid peptide 1-28 (Abeta(28)) and the resulting {Ru(CO)(3)}(2+) peptide adduct was investigated by a variety of biophysical methods. (1)H NMR titrations highlighted a selective interaction of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service