주요 문서
796-0659
HPLC Analysis of Human Growth Hormone (hGH) on Discovery® BIO PolyMA-WAX
application for HPLC
로그인조직 및 계약 가격 보기
About This Item
기술
HPLC: suitable
검사 파라미터
column: Discovery BIO PolyMA-WAX, 5 cm × 4.6 mm I.D., 5 μm particles (59602-U)
mobile phase: [A] 20 mM Tris-HCl, pH 8.0; [B] 20 mM Tris-HCl, 0.5 M NaCl, pH 8.0
gradient: 5 to 70% B in 30 min. (linear)
flow rate: 0.5 mL/min
column temp.: 25 °C
detector: UV, 210 nm
injection: 10 mg hGH
적합성
application for HPLC
응용 분야
life science and biopharma
분석 메모
This Figure shows the degradation products of human growth hormone (hGH) well-separated on a Discovery BIO PolyMA-WAX anion-exchange column. The difference between the resolved compounds is the conversion of protein amide(s) to carboxylate(s), demonstrating both the power of the ion-exchange technique, and the efficiency of the Discovery BIO PolyMA-WAX column.
법적 정보
Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany
분석물
설명
    Human Growth Hormone human, HGH, recombinant, expressed in HEK 293 cells, HumanKine®, suitable for cell culture
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)