주요 문서
57634
Stator Face Assembly
for use with model 7725 injector
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41115854
호환성
for use with model 7725 injector
일반 설명
Stator Face Assembly is an accessory for use with Rheodyne® model 7725 injector employed for the introduction of the analyte into high performance liquid chromatograph prior to analytical determinations.
추천 제품
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
법적 정보
Rheodyne is a registered trademark of IDEX Health & Science, LLC
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)