주요 문서
57621
Rheodyne® Injector
Rheodyne 7725i
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41115854
설명
internal position sensing switch
특징
manual
number of ports 6
positions 2
제조업체/상표
Rheodyne 7725i
파라미터
7000 psi max. pressure
일반 설명
Rheodyne® Injector model is a stainless steel version and comprises of a six-port sample injection valves in which the sample is loaded through a built-in needle port in the front of the valve.
추천 제품
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
법적 정보
Rheodyne is a registered trademark of IDEX Health & Science, LLC
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)