주요 문서
22357
Chart Paper for Recorder
box of 4
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
14111500
특징
chart type blank roll
포장
box of 4
제조업체/상표
Agilent/HP 5181-1219 (OEM)
호환성
for use with Agilent/HP 3394,3395, 3396
유사한 제품을 찾으십니까? 방문 제품 비교 안내
애플리케이션
Analog signal from the chromatograph may be connected to a recorder with chart paper speed of 5 min/in. during an analysis to measure Glutethimide concentrations in the serum and cerebrospinal fluid (CSF) using gas chromatography.
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)
문서 라이브러리 방문
John Kenkel
Analytical Chemistry for Technicians, 366-366 (1994)
A.V.Bakshi U.A.Bakshi
Electronic Measurements & Instrumentation (2008)
Comparison of glutethimide concentration in the serum and cerebrospinal fluid of humans in drug overdose.
M Gold et al.
Clinical chemistry, 19(10), 1158-1161 (1973-10-01)