주요 문서
Z742622
Centrifuge Tube Adapter For Tube-O-DIALYZER
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41123100
NACRES:
NB.78
포장
pkg of 2 ea
제조업체/상표
G-BioSciences 786-145
일반 설명
The Centrifuge Tube Adaptor for Tube-O-Dialyzer is designed to assist centrifugation of Tube-O-Dialyzer using a bench top centrifuge. The adaptor can be used for both Micro and Medi size Tube-O-Dialyzer. The Tube-O-Dialyzer can be positioned into the adaptor either upright position or inverted position for centrifugation.
법적 정보
Tube-O-DIALYZER is a trademark of Geno Technology, Inc.
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)