주요 문서
Z692697
microDOC Compact gel documentation system, with UV Transilluminator (UVT254)
AC/DC input 110 V AC
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41105300
NACRES:
NB.22
제조업체/상표
Cleaver CSL-MDOCUV254$
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
This option combines the microDOC Compact Gel Documentation System with a 12 x 12 cm UV 254 nm tranilluminator.
Available with optional TotalLab 1D software. Main software functions include: lane creation, background subtraction, band detection, profile deconvolution, and molecular size, pI and quantity calibration.
System comprises of:
Canon G12 digital
2MB storage card
Ethidium bromide & close up +3 filters
Lift-off hood with easy access door and built-in inner lights
8 in. TFT color monitor
12 x 12 cm UV 254 nm transilluminator
법적 정보
TotalLab is a trademark of Nonlinear Dynamics Ltd.
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)