Key Documents
Z616850
Vacuum controller for Buchi® evaporators and vacuum pumps
V-850, output230 V
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41105101
포장
pack of 1 ea
제조업체/상표
Büchi 47299
재고 정보
available only in EU
출력
230 V
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
Ideally suitable for the V-700/710 vacuum pumps, but is compatible with all older models. When used with the pump or the Rotavapor, a separate power cord is not required. However, if the vacuum controller is to be used as a stand-alone device it will require a power cord.
V-850 features:
  • Regulation to a specified setpoint
  • Library with 43 pre-determined solvents
  • Timer function
  • Control of jack and rotation on the Rotavapor
V-855 features:
  • EasyVac function when used with V-700/710 vacuum pump provides automatic process control based on vapor pressure detection
  • Automatic distillation with the unique stage probe
  • Programming for pressure gradients in specialized distillation
  • Repeat function for reproducible repetition of a distillation
법적 정보
Büchi is a registered trademark of Büchi Labortechnik AG