주요 문서
HS1040A
Clearly Safe® glasses dispenser, counter or wall mountable
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41121800
NACRES:
NB.51
material
blue acrylic
제조업체/상표
Heathrow Scientific HS1040A
일반 설명
Ergonomically angled for easy dispensing this translucent acrylic dispensers provide easy access to safety accessories. The hinged lid protects glasses from dust and debris. Can be mounted on counter or wall.
특징 및 장점
  • Hinged lid protects glasses from dust and debris
  • Set on counter or wall mount (screws included)
  • Large dispenser holds approximately 20 glasses
  • Small dispenser holds approximately 10 glasses
  • Great for holding other personal items
법적 정보
Clearly Safe is a registered trademark of Heathrow Scientific LLC
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)