주요 문서
GE18-1004-30
Immobiline® DryStrip kit, for Immobiline DryStrip gels with MultiPhor II Electrophoresis System
pack of 1 kit
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41105300
NACRES:
NB.22
특징
wetted part: no
포장
pack of 1 kit
제조업체/상표
Cytiva 18100430
일반 설명
This kit is designed for running the first dimension of 2-D electrophoresis using precast Immobiline DryStrip gels, see Immobiline DryStrip Gels. You can apply up to 1NANA µl of sample per strip and run up to 12 strips simultaneously
This Instruction manual presents all necessary information for the entire method for horizontal 2-D electrophoresis. By appropriate adaptations, the SDS electrophoresis can also be performed using vertical electrophoresis equipment. Isoelectric focusing with an immobilized pH gradient (IPG), makes true isoelectric focusing possible and significantly improves the reproducibility of the spot distribution along the pH gradient axis of 2-D maps. IPG also makes it possible to focus basic proteins in the gel and to obtain distinct protein spots. Immobiline DryStrip kit facilitates sample application and running and equilibration of Immobiline DryStrip for the first dimension in 2-D (two-dimensional) electrophoresis
특징 및 장점
  • For use of immobiline DryStrip gels with Multiphor II electrophoresis system.
법적 정보
Immobiline is a registered trademark of Cytiva
MultiPhor is a trademark of Cytiva
Storage Class Code
13 - Non Combustible Solids
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)
문서
This page describes troubleshooting first-dimension isoelectric focusing using MultiPhor™ II Electrophoresis System and Immobiline® DryStrip Kit.