Key Documents
28-3690
Sigma-Aldrich
Sodium persulfate
SAJ first grade, ≥98.0%
동의어(들):
Sodium peroxodisulfate
로그인조직 및 계약 가격 보기
About This Item
Linear Formula:
Na2S2O8
CAS Number:
Molecular Weight:
238.10
EC Number:
MDL number:
UNSPSC 코드:
12352302
PubChem Substance ID:
Grade
SAJ first grade
분석
≥98.0%
형태
fine crystals
반응 적합성
reagent type: oxidant
재고 정보
available only in Japan
pH
2.5-4.0
SMILES string
[Na+].[Na+].[O-]S(=O)(=O)OOS([O-])(=O)=O
InChI
1S/2Na.H2O8S2/c;;1-9(2,3)7-8-10(4,5)6/h;;(H,1,2,3)(H,4,5,6)/q2*+1;/p-2
InChI key
CHQMHPLRPQMAMX-UHFFFAOYSA-L
유사한 제품을 찾으십니까? 방문 제품 비교 안내
픽토그램
Flame over circleHealth hazardExclamation mark
신호어
Danger
유해 및 위험 성명서
Hazard Classifications
Acute Tox. 4 Oral - Ox. Sol. 3 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3
표적 기관
Respiratory system
Storage Class Code
5.1B - Oxidizing hazardous materials
WGK
WGK 1
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
개인 보호 장비
Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges
문서 라이브러리 방문
Anhua Long et al.
Environmental science and pollution research international, 22(15), 11606-11616 (2015-04-08)
An electro/Fe(2+)/persulfate process has been conducted for toluene removal from surfactant (SDS) flushing solution, and the pseudo-second-order reaction rate constant (k2 value) of toluene removal has been optimized by a response surface methodology (RSM). The results indicated that in this
N Sabri et al.
The Science of the total environment, 427-428, 382-389 (2012-05-15)
The objective of this work was to evaluate the removal of ibuprofen (IBP) using the oxidants hydrogen peroxide (H(2)O(2)) and sodium persulfate (Na(2)S(2)O(8)). The ability of magnetite (Fe(3)O(4)) to activate persulfate (PS) and H(2)O(2) for the oxidation of IBP at
Amy L Teel et al.
Journal of hazardous materials, 196, 153-159 (2011-10-05)
The potential for 13 naturally occurring minerals to mediate the decomposition of persulfate and generate a range of reactive oxygen species was investigated to provide fundamental information on activation mechanisms when persulfate is used for in situ chemical oxidation (ISCO).
Mushtaque Ahmad et al.
Journal of contaminant hydrology, 115(1-4), 34-45 (2010-05-05)
Persulfate dynamics in the presence of subsurface minerals was investigated as a basis for understanding persulfate activation for in situ chemical oxidation (ISCO). The mineral-mediated decomposition of persulfate and generation of oxidants and reductants was investigated with four iron and
Aikaterini Tsitonaki et al.
Water research, 42(4-5), 1013-1022 (2007-10-19)
The effects of heat-activated persulfate on indigenous microorganisms and microcosms augmented with Pseudomonas putida KT2440 were studied in laboratory batch reactors with aquifer material. Microscopic enumeration was used to measure the changes in cell density, and acetate consumption was used