주요 문서
Z743110
Julabo® flow through immersion cooler
FD 200, temp. range 10 °C (30 °C), AC/DC input 230 V AC, UK plug
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41103706
NACRES:
NB.77
포장
pack of 1 ea
제조업체/상표
FD 200
Julabo 9655825
파라미터
10 °C temp. range (30 °C)
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
JULABO flow-through coolers are the perfect complement to open bath circulators, immersion circulators and bridge mounted circulators extending their temperature range to below ambient temperature. They offer a small footpri nt, have a low energy requirement and represent an economic solution preventing tap water usage for cooling. Instruments are designed for integration into an existing cooling loop (e.g. consisting of open bath circulator wit h pump).
Your advantages
  • For applications near ambient temperatures
  • Integrated freezing protection
  • Preventing the use of precious tap water for cooling
  • Compact design, small footprint
  • Ease of operation
  • Low energy requirement
  • Time saving - Comfortable and simple operation for setpoint adjustment
  • Precise Temperature Control - PID Temperature control with set control parameters, temperature stability ±0.02...±0.2 °C
법적 정보
Julabo is a registered trademark of Julabo USA, Inc.
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)