주요 문서
Z220590
Sigma-Aldrich
Solid-transfer vessel with two side-arms and a gas inlet with glass stopcock
capacity 75 mL, side arm male Joints: ST/NS 14/20 (2)
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41121815
NACRES:
NB.43
라벨
U
조인트
male Joints: ST/NS side arm14/20 (2)
male joint ST/NS top45/40
용량
75 mL
일반 설명
For transferring and packaging air-sensitive solid compounds under inert atmosphere.
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)
문서
Versatile system for chemical reactions, extractions, and transfers of solids and liquids in laboratories.