주요 문서
DWK73750-16150
KIMBLE® disposable screw thread culture tube with white marking spot
round bottom borosilicate glass, tube diam. × L (16 mm) (105 mm), non-sterile
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41121706
NACRES:
NB.11
material
round bottom borosilicate glass
round bottom
screw top neck
transparent borosilicate glass tube
무균
non-sterile
특징
without cap
포장
case of 1000 ea Modular tray shrink-wrapped in plastic film
제조업체/상표
Kimble® 73750-16150
튜브 직경 × 길이
(16 mm) (105 mm)
튜브 직경
(16 mm)
피팅
neck (External Thread)
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
Disposable Screw Thread Culture Tubes with Marking Spot
  • These culture tubes are suitable for tissue culture work and general bacteriological use.
  • Round-bottomed
  • Each tube has a chevron marking area
  • Supplied without caps
  • Manufactured from 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements
특징 및 장점
  • Manufactured from borosilicate glass 5.1
  • Round-bottomed
  • Each tube has a chevron marking area
  • Modular trays are shrink-wrapped in plastic film
  • Supplied without caps
법적 정보
KIMBLE is a registered trademark of DWK Life Sciences
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)