주요 문서
BAF171310024
SP Bel-Art® Pipette Rinser for Cleanware Pipette Rinsing System
diam. × H 25.2 cm (9.9) × 79 cm (31.25)
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41122800
NACRES:
NB.01
특징
to hold For 61 cm (24") pipettes
포장
pack of 1 ea
제조업체/상표
Bel-Art F17131-0024
직경 × 높이
25.2 cm (9.9) × 79 cm (31.25)
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
The Cleanware Pipette Rinsing System for pipette cleaning and storage has an effective, reliable design that ensures safe, efficient operation. Available in a complete system or in individual components.
특징 및 장점
  • Pipette Rinsers provides a simple, automatic method for rinsing large numbers of pipettes at one time
  • High level water entry provides an air gap between the maximum water level and the end of the water inlet tube ensuring safe and efficient operation
  • “Tube within a tube” siphoning system assures rapid, complete and automatic siphoning and refilling over a wide range of inlet water pressures and flow rates
  • Minimizes the danger of siphon stalling at certain water inflow rates and prevents any possible backflow into the potable water supply in the event of a pressure interruption
  • Leak-proof, polyethylene Pipette Jars store and soak pipettes and will withstand all common cleaning solutions at room temperature, including sulfuric acid/dichromate
  • Pipette Baskets, filled with pipettes, fit into the corresponding size jar; cut-out hand grips for easy handling; gusher holes provide quick drainage when lifted from a soaking jar; foam disk on the bottom cushions pipette tips to prevent breakage
  • Includes 30.5cm (12”) long piece of 2.5cm (1”) I.D. drain tube and 1.5M (5ft) of 12.5mm (½”) I.D. flexible water inlet tube
법적 정보
Bel-Art is a registered trademark of SP Scienceware, a division of SP
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)