Merck
Rogard II Prefilter 30 cm (12 in.), Pretreats feed water to protect the reverse osmosis (RO) membrane
Rogard II Prefilter
Rogard II Prefilter 30 cm (12 in.), Pretreats feed water to protect the reverse osmosis (RO) membrane
Products
製品区分
ブランド
品質レベル
孔径(µm)