コンテンツへスキップ
MilliporeSigma
給水圧調整器 Regulates the feed water pressure from 0-25 bars, Ensures accurate water level sensing to prevent overflow for use with high flow water systems
給水圧調整器
給水圧調整器 Regulates the feed water pressure from 0-25 bars, Ensures accurate water level sensing to prevent overflow for use with high flow water systems
Products
製品区分
ブランド
長さ(mm)
適合性