Merck
Carbon Cartridge (4" x 20"), For pretreatment of water with high chlorine level (> 3 ppm)
Carbon Cartridge
Carbon Cartridge (4" x 20"), For pretreatment of water with high chlorine level (> 3 ppm)