Dual-Labeled Probes
- Amounts: 1, 3, 5, and 10 OD
- Purification: HPLC
- Sequence Lengths: 15 - 40 bases
- Modifications: Locked Nucleic Acid available
- Dyes: Comprehensive offering of reporters and quenchers
- Quality Control: 100% mass spectrometry
- Format: Supplied dry in amber tubes
- Learn more
Molecular Beacons
- Amounts: 1, 3, 5, and 10 OD
- Purification: HPLC
- Sequence Lengths: 15 - 40 bases
- Modifications: Locked Nucleic Acid available
- Dyes: Comprehensive offering of reporters and quenchers
- Quality Control: 100% mass spectrometry
- Format: Supplied dry in amber tubes
- Learn more
LightCycler® Probes
- Amounts: 0.1, 0.25, 1.5, and 15 OD
- Purification: HPLC
- Sequence Lengths: 15 - 40 bases
- Modifications: Locked Nucleic Acid available
- Dyes: Probe 1 fluorescein (3′ end); Probe 2 phosphate (3′ end) and LC Red 610, 640, 670 or 705 (5′ end)
- Quality Control: 100% mass spectrometry
- Format: Supplied dry in amber tubes
Scorpions® Probes
- Amounts: 1, 5, and 5 OD
- Purification: HPLC
- Sequence Lengths: 30 - 60 bases
- Modifications: Locked Nucleic Acid available
- Dyes: Comprehensive offering of reporters and quenchers
- Quality Control: 100% mass spectrometry
- Format: Supplied dry in amber tubes
Locked Nucleic Acid
- Available for all Custom qPCR Probe Types
- Increased thermal stability and hybridization specificity
- Easier and more specific designs for problematic target sequences
- Learn more
Black Hole Quencher® (BHQ®)
- Available for all Custom qPCR Probe types
- No native fluorescence (emits heat instead of light), resulting in lower background fluorescence
- Increased signal-to-noise ratio, providing higher sensitivity
- Enables a wider choice of reporters for multiplexing
- Learn more