Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

43954

Supelco

L-3-Aminoisobutyric acid

analytical standard

Synonym(s):

(+)-β-Aminoisobutyric acid, (2S)-3-Amino-2-methylpropanoic acid, (S)-β-AIB, (S)-β2-Homoalanine, (S)-3-Amino-2-methylpropionic acid, L-α-Methyl-β-alanine, S-BAIBA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H9NO2
CAS Number:
Molecular Weight:
103.12
MDL number:
UNSPSC Code:
41116107

grade

analytical standard

Assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

clinical testing

format

neat

SMILES string

C[C@@H](CN)C(O)=O

InChI

1S/C4H9NO2/c1-3(2-5)4(6)7/h3H,2,5H2,1H3,(H,6,7)/t3-/m0/s1

InChI key

QCHPKSFMDHPSNR-VKHMYHEASA-N

Looking for similar products? Visit Product Comparison Guide

General description

L-3-Aminoisobutyric acid, an enantiomer of β-aminoisobutyric acid (BAIBA), is derived from valine in the mitochondria of primarily skeletal muscle.

Application

L-3-Aminoisobutyric acid may be used as an analytical reference standard for the quantification of the analyte in biological fluids using dual column reversed-phase HPLC-fluorescence technique.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

43954-BULK:
43954-10MG:
43954-50MG:
43954-VAR:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service