Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

42810

Supelco

2-Aminoethyl diphenylborinate

for TLC derivatization, ≥97.0%

Synonym(s):

2-APB, 2-Aminoethoxydiphenyl borate, 2-Aminoethyl diphenyl borate, Diphenylboric acid 2-aminoethyl ester, Diphenylborinic acid, NSC 17107

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2BOCH2CH2NH2
CAS Number:
Molecular Weight:
225.09
Beilstein:
2942273
EC Number:
MDL number:
UNSPSC Code:
12164500
PubChem Substance ID:

grade

for TLC derivatization

Assay

≥97.0% (NT)
≥97.0%

application(s)

general analytical

SMILES string

NCCOB(c1ccccc1)c2ccccc2

InChI

1S/C14H16BNO/c16-11-12-17-15(13-7-3-1-4-8-13)14-9-5-2-6-10-14/h1-10H,11-12,16H2

InChI key

BLZVCIGGICSWIG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Reagent for the densitometric determination of flavonoids

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

42810-1G:
42810-5G:
42810-25G:
42810-VAR:
42810-BULK:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T. Brasseur et al.
Journal of Chromatography A, 351, 351-351 (1986)
Evaluation of the antioxidant activity of asparagus, broccoli and their juices.
Sun, Ting, Joseph R. Powers, and Juming Tang
Food Chemistry, 105, 101-106 (2007)
M. Billeter et al.
J. Planar Chromatogr., 3, 370-370 (1990)
Jun-Ichi Goto et al.
Cell calcium, 47(1), 1-10 (2009-12-01)
Store-operated calcium entry (SOCE) or calcium release-activated calcium current (I(CRAC)) is a critical pathway to replenish intracellular calcium stores, and plays indispensable roles in cellular functions such as antigen-induced T lymphocyte activation. Despite the importance of I(CRAC) in cellular functions
[Densitometric analysis of heterosides of quercetin in plant extracts].
T Brasseur et al.
Journal of chromatography, 437(1), 260-264 (1988-03-11)

Protocols

Qualitative Thin Layer Chromatography Analysis of Flavonoids and Quantification of Terpene Lactones in Ginkgo Biloba Extracts and Tablets

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service