Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

DF-001-B

Sigma-Aldrich

Custom - DMEM/F12 (50:50) w/ NEAA, 500ml

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200

Quality Level

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

DF-001-B:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service