Skip to Content
Merck
All Photos(3)

Key Documents

Safety Information

07-1590

Sigma-Aldrich

Anti-phospho Histone H2A.X Antibody (Tyr142)

from rabbit, purified by affinity chromatography

Synonym(s):

Histone H2A.x, H2a/x

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse

technique(s)

dot blot: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

Gene Information

human ... H2AX(3014)
mouse ... H2Ax(15270)
rat ... H2Ax(500987)

General description

Histone H2A is one of the 5 main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N terminal tail H2A is involved with the structure of the nucleosomes of the ′beads on a string′ structure. The phosphorylated form of H2A.X is thought to be invloved with DNA repair, possibly through reorganizing chromatin into a more open state and allowing the recruitment of proteins to repair DNA double stranded breaks.

Immunogen

KLH-conjugated linear peptide corresponding to the C-terminus of human phospho Histone H2A.X phosphorylated at Tyr142.

Application

Anti-phospho Histone H2A.X Antibody (Tyr142) is a highly specific rabbit polyclonal antibody, that targets Histone H2A.X & has been tested in western blotting & Dot Blot.
Dot Blot Analysis: 0.5 µg/mL from a representative lot detected phospho Histone H2A.X (Tyr142) peptides, but not phospho Histone H2A.X (Tyr142) peptides or other peptides corresponding to modified histones.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones

Quality

Evaluated by Western Blotting in colcemid treated HeLa cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected phospho Histone H2A.X (Tyr142) in colcemid treated HeLa cell lysate.

Target description

~16 kDa observed. Uncharacterized band(s) may be observed in some cell lysates.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

07-1590:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prachi Borude et al.
Gene expression, 18(2), 115-123 (2018-03-16)
Acetaminophen (APAP) overdose is the leading cause of acute liver failure (ALF) with limited treatment options. It is known that liver regeneration following APAP-induced ALF is a deciding factor in the final outcome. Previous studies from our laboratory using an
I M Wilson et al.
Oncogene, 33(36), 4464-4473 (2013-10-08)
In an effort to identify novel biallelically inactivated tumor suppressor genes (TSGs) in sporadic invasive and preinvasive non-small-cell lung cancer (NSCLC) genomes, we applied a comprehensive integrated multiple 'omics' approach to investigate patient-matched, paired NSCLC tumor and non-malignant parenchymal tissues.
WSTF regulates the H2A.X DNA damage response via a novel tyrosine kinase activity.
Xiao, A; Li, H; Shechter, D; Ahn, SH; Fabrizio, LA; Erdjument-Bromage, H; Ishibe-Murakami et al.
Nature null
Zu Ye et al.
Science advances, 7(32) (2021-08-06)
DNA double-strand break (DSB) repair is initiated by MRE11 nuclease for both homology-directed repair (HDR) and alternative end joining (Alt-EJ). Here, we found that GRB2, crucial to timely proliferative RAS/MAPK pathway activation, unexpectedly forms a biophysically validated GRB2-MRE11 (GM) complex
MDC1 cleavage by caspase-3: a novel mechanism for inactivating the DNA damage response during apoptosis.
Solier, S; Pommier, Y
Cancer Research null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service