Skip to Content
Merck
All Photos(2)

Key Documents

810157C

Avanti

14:0 Liss Rhod PE

Avanti Research - A Croda Brand 810157C

Synonym(s):

1,2-dimyristoyl-sn-glycero-3-phosphoethanolamine-N-(lissamine rhodamine B sulfonyl) (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C60H97N4O14PS2
CAS Number:
Molecular Weight:
1193.53
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (810157C-1mg)
pkg of 5 × 1 mL (810157C-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810157C

concentration

1 mg/mL (810157C-1mg)
1 mg/mL (810157C-5mg)

shipped in

dry ice

storage temp.

−20°C

Application

14:0 Liss Rhod PE or 1,2-dimyristoyl-sn-glycero-3-phosphoethanolamine-N-(lissamine rhodamine B sulfonyl) (ammonium salt) has been used in the preparation of nanoemulsion to visualize its cellular uptake by microscopy. 14:0 Liss Rhod PE is also suitable for the preparation of: labeled phospholipid vesicle for lipid mixing assay, supported lipid bilayer (SLB) vesicles for surface plasmon resonance (SPR)-binding experiments, red labeled liposome

Packaging

5 mL Amber Glass Screw Cap Vial (810157C-1mg)
5 mL Amber Glass Screw Cap Vial (810157C-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oil-in-water nanoemulsions are suitable for carrying hydrophobic compounds: Indomethacin as a model of anti-inflammatory drug
Kwasigroch B, et al.
The International journal of pharmacy practice null
Induction of differential T-cell epitope by plain-and liposome-coupled antigen
Taneichi M, et al.
Bioconjugate Chemistry, 8(23) (2018)
Noel Vera-González et al.
Journal of biomedical materials research. Part A, 108(11), 2263-2276 (2020-05-05)
Fungal infections can cause significant patient morbidity and mortality. Nanoparticle therapeutics have the potential to improve treatment of these infections. Here we report the development of liposomal nanoparticles incorporating anidulafungin, a potent antifungal, with the goal of increasing its solubility
Nicholas L Angeloni et al.
Scientific reports, 6, 22915-22915 (2016-03-12)
Exosomes are produced by cells to mediate intercellular communication, and have been shown to perpetuate diseases, including cancer. New tools are needed to understand exosome biology, detect exosomes from specific cell types in complex biological media, and to modify exosomes.
Juan J Domínguez Pardo et al.
Biophysical journal, 115(1), 129-138 (2018-07-05)
Styrene-maleic acid (SMA) copolymers have attracted interest in membrane research because they allow the solubilization and purification of membrane-spanning proteins from biological membranes in the form of native-like nanodisks. However, our understanding of the underlying SMA-lipid interactions is hampered by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service