Merck
HPLC Analysis of Beta-Receptor Agonist Enantiomers on Astec® CHIROBIOTIC® T
HPLC Analysis of Beta-Receptor Agonist Enantiomers on Astec® CHIROBIOTIC® T application for HPLC
Materiali
Colonna analitica
N° Catalogo
Descrizione
Determinazione del prezzo
Colonna HPLC chirale Astec® CHIROBIOTIC® T
5 μm particle size, L × I.D. 25 cm × 4.6 mm
Standard
N° Catalogo
Descrizione
Determinazione del prezzo
Clenbuterol hydrochloride
≥95%
(S)-(−)-Atenolol
powder
(±)-Sotalol hydrochloride
≥98% (TLC), powder
Atenolol
≥98% (TLC), powder
(±)-Metoprolol (+)-tartrate salt
≥98% (titration), powder
(R)-(+)-Atenolol
99%
(S)-(−)-Atenolol
99%
Atenolol-d7
analytical standard
CONDITIONS
column
CHIROBIOTIC T, 25 cm x 4.6 mm, 5 μm particles (12024AST)
mobile phase
15 mM ammonium formate in methanol
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 220 nm