GC Analysis of α-Hexachlorocyclohexane (α-HCH) Enantiomers on Astec® CHIRALDEX™ G-DP
![GC Analysis of α-Hexachlorocyclohexane (α-HCH) Enantiomers on Astec® CHIRALDEX™ G-DP suitable for GC](/deepweb/assets/sigmaaldrich/product/images/170/366/56e04461-2f1e-47ca-9536-6746afa53b9d/800/56e04461-2f1e-47ca-9536-6746afa53b9d.jpg)
Materiali
Colonna analitica
column
Astec CHIRALDEX G-DP, 30 m x 0.25 mm I.D., 0.12 μm (78033AST)
oven
170 °C
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 30 psi
injection
0.2 μL, 80:1 split
sample
2 mg/mL in ethanol
peaks 1 & 2: α-HCH (α-hexachlorocyclohexane) enantiomers