G005061
GC Analysis of Methionine Methyl Ester Enantiomers (N-Trifluoroacetyl Derivatives) on Astec® CHIRALDEX™ G-TA
suitable for GC
About This Item
tecniche
gas chromatography (GC): suitable
Parametri analitici
column: Astec CHIRALDEX G-TA, 30 m x 0.25 mm I.D., 0.12 μm (73033AST)
oven: 160 °C
inj. temp.: 250 °C
detector: FID, 250 °C
carrier gas: helium, 30 psi
sample: peaks 1 & 2: methionine methyl ester enantiomers (N-TFA derivatives)
applicazioni
food and beverages
life science and biopharma
Note legali
Colonna analitica
Standard
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)