Documenti fondamentali
509590
Syringes Fitted with Luer Lock Valve (509353)
volume 1000 μL, 1MDR-VLL-GT, needle size (not included)
About This Item
Codice UNSPSC:
41122001
Materiali
glass syringe
Misura ago
(not included)
Caratteristiche
Pre-fitted with Luer Lock Valve
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
SGE 008160
Volume
1000 μL
Descrizione generale
  • SGE Syringes (volume 1 mL, needle size (not included)) fitted with Luer Lock Valve, Model: 1MDR-VLL-GT.
  • Made using borosilicate glass, PTFE plunger tips, and KEL-F Luer cones.
  • Autoclavable syringes but the plunger needs to be removed.
  • Feature an interference fit between the plunger tip and the barrel, that improves the gas sealing properties of the syringe.
  • Maximum inertness for liquid or gas samples.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
I clienti hanno visto anche
Global Trade Item Number
SKUGTIN
5095904061833037690