Documenti fondamentali
22478
Supelco
Boccole Supeltex M-4 per colonne impaccate, D.E. colonna 1/4”
I.D. 1/4 in., 1/4 in. O.D. Column, pkg of 50 ea
About This Item
Codice UNSPSC:
41121815
Confezionamento
pkg of 50 ea
Diam. int.
1/4 in.
Raccordi
1/4 in. GC fitting ferrule for 1/4 in. O.D. packed column
Compatibilità
1/4 in. O.D. Column
Descrizione generale
Supelco has refined the design of these graphite ferrules to provide a clean, sharp profile with minimal flash.
Characteristics:
Seals at one-quarter turn past fingertight. Maximum sealing surface contact, reduced risk of column contamination at installation.
Material Composition: flexible graphite
Maximum Temperature: 450 °C
Note legali
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)