Z742780
smartPans™ cahn style aluminim weighing dish
pack of 100 (Dishes supplied in flip top vial)
About This Item
Codice UNSPSC:
41121812
NACRES:
NB.35
Prodotti consigliati
Materiali
aluminum
Confezionamento
pack of 100 (Dishes supplied in flip top vial)
Produttore/marchio commerciale
LevGo Inc
Diam. × Alt. × spessore
25 mm × 3.59 mm × 0.038 mm
Peso
53 mgEach boat
Descrizione generale
Ideal for microbalances, organic micro elementary / thermal analysis, and pharmaceutical use.
- Made of Ultra High Vacuum (UVH) Foil - “A” wettable and completely free from oils normally found on other grades of aluminum foil.
- Foil meets ASTM B-479, specification 3.1.4 and 10.3.1.
- “Cahn style” accessory pan / disc format
- Compatible with most commercially available analyzers and microbalances
- Foil and Pans are made in the USA
- Packaged 20 or 100 per easy-to-use PP flip top vials - Both the pans and the vials recyclable
Note legali
smartPans is a trademark of LevGo Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)