Documenti fondamentali
Z637548
BRAND® PP funnels, plain interior
top diam. 50 mm, stem O.D. × L 9 mm × 43 mm
About This Item
Codice UNSPSC:
41121809
NACRES:
NB.43
Produttore/marchio commerciale
BRAND 147015
D.E. stelo × lungh.
9 mm × 43 mm
Diametro sommità
50 mm
Descrizione generale
Made from durable polypropylene (PP), these funnels offer excellent chemical resistance and are suitable for a wide range of applications. The plain interior facilitates easy cleaning and prevents contamination, ensuring reliable performance during use.
Altre note
BR147015 (India)
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)