Key Documents
Z330264
BRAND® dropping bottle, LDPE
capacity 100 mL
Sinonimo/i:
dropping bottles
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.17
Materiali
low-density polyethylene bottle
narrow-mouth bottle
round bottle
translucent bottle
Caratteristiche
with cap
Confezionamento
pkg of 50
Produttore/marchio commerciale
BRAND 125238
Capacità
100 mL
diam. × alt.
45 mm × 136 mm
Raccordi
GL 18 thread
Descrizione generale
Flexible. Detachable threaded closure with dropper nozzle and attached spout cap.
Altre note
BR125238 (India)
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Raccomandato