Documenti fondamentali
Z327034
BRAND® BLAUBRAND® volumetric flask, glass stopper, amber glass
volume 25 mL, accuracy: 0.04 mL
About This Item
Codice UNSPSC:
41121804
NACRES:
NB.43
Materiali
borosilicate glass
Giunto
joint ST/NS neck10/19
Sterilità
non-sterile
Caratteristiche
class A DE-M, DIN EN ISO 1042. Calibrated to contain (TC, In).
closure type Glass stopper
Confezionamento
pack of 2
Produttore/marchio commerciale
BRAND
Campo di misura del volume
25 mL, accuracy: 0.04 mL
Capacità
25 mL
Descrizione generale
Volumetric flasks according to DIN EN ISO 1042, Class A, conformity certified. Made from Boro 3.3, calibrated to contain (TC, IN). Supplied with a interchangeable amber glass stopper. Marks and inscriptions in high contrast white enamel.
All BLAUBRAND® volumetric flasks are supplied with a batch number and an accompanying batch certificate in the original packaging.
Altre note
BR37447 (India)
Note legali
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
Protocolli
This page contains detailed information on working with volumetric flasks and how to prepare a standard solution with a volumetric flask.