Key Documents
TMO312087-0004
Nalgene® narrow-mouth bottle
natural polypropylene copolymer, size 125 mL, case of 500 ea
Sinonimo/i:
Nalgene bottle, plastic bottle
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.17
Materiali
natural polypropylene copolymer
Confezionamento
case of 500 ea
Produttore/marchio commerciale
Nalgene 312087-0004
Misura
125 mL
Descrizione generale
Store and ship light-sensitive reagents with Thermo Scientific Nalgene Narrow-Mouth Opaque Amber HDPE Packaging Bottles with Closure. Ideal for diagnostics, biologicals, specialty chemicals, reagents, adhesives and veterinary pharmaceuticals, these high density polyethylene bottles are injection blow-molded for uniform wall thickness and strength. They meet USP light transmission standards and are bulk packed for packaging applications.
Note legali
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries