Key Documents
MTCP3801QF
MTC Bio Pure.AmpqPCR 8 Tube Strips
capacity 0.1 mL, tube frosted, separate, optical clear flat caps
Sinonimo/i:
PCR sample storage tubes, plastic tube, sample storage tube
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.15
Materiali
clear cap (optical)
clear tube
polypropylene
Sterilità
non-sterile
Confezionamento
pack of 120 ea
Produttore/marchio commerciale
MTC Bio P3801-QF
Capacità
0.1 mL
Descrizione generale
0.1ml qPCR 8-Strip (With SEPARATE Optical Strip Caps) FROSTED, 120/pk
Pure.Amp 0.1mL qPCR strips strips are precision manufactured to maintain tight dimensional tolerances of ultra-thin wall thickness, for improved thermal transfer and are certified free of RNase, DNase, DNA and PCR inhibitors, and are made of heat-resistant medical-grade polypropylene.
Note legali
MTC is a trademark of MTC Bio
Pure.Amp is a trademark of MTC Bio