Documenti fondamentali
MTCH6800200
MTC Bio Halo Lightweight Single Channel Pipettor
capacity 20-200 μL
Sinonimo/i:
pipette
About This Item
Codice UNSPSC:
41121500
NACRES:
NB.15
Caratteristiche
adjustable
manual
no. of channels 1
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
MTC Bio H6800-200
Capacità
20-200 μL
Categorie correlate
Descrizione generale
As one of the lightest pipettors ever introduced, the Halo Pipettor dramatically reduces the risk of repetitive stress injury and the general shoulder, arm, and hand fatigue associated with daily pipetting. Reduced actuation forces andan ergonomic design make Halo Pipettors feel balanced and natural in hand. Featuring a four-digit micrometer display for precise volume selection and a click-based adjustment wheel with DriftGuard protection, the Halo Pipettor is perfect for applications that demand exceptional accuracy. With a recessed ejector button, and cushioned, shock-absorbing stop, the ergonomics of tip ejection have been re-imagined to relieve stress on the thumb and joints.
Halo Single Channel Pipettor, lightweight, 200-1250µL
Caratteristiche e vantaggi
  • 10-40% lighter than leading competitors
  • 4 Digit micrometer with DriftGuard protection
  • Exclusive 1250µL model for extended length tips
  • User-enabled calibration port
  • Compatible with universal tips
Note legali
Halo is a trademark of MTC Bio
MTC is a trademark of MTC Bio
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
Global Trade Item Number
SKUGTIN
MTCH6800200-1EA4065271080409