MTCH680010M
MTC™ Bio Halo™ Lightweight Single Channel Pipettor
capacity 1-100 μL
Sinonimo/i:
pipette
About This Item
Codice UNSPSC:
41121500
NACRES:
NB.15
Prodotti consigliati
Caratteristiche
adjustable
manual
no. of channels 1
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
MTC™ Bio H6800-10M
Capacità
1-100 μL
Descrizione generale
As one of the lightest pipettors ever introduced, the Halo™ Pipettor dramatically reduces the risk of repetitive stress injury and the general shoulder, arm, and hand fatigue associated with daily pipetting. Reduced actuation forces andan ergonomic design make Halo™ Pipettors feel balanced and natural in hand. Featuring a four-digit micrometer display for precise volume selection and a click-based adjustment wheel with DriftGuard protection, the Halo™ Pipettor is perfect for applications that demand exceptional accuracy. With a recessed ejector button, and cushioned, shock-absorbing stop, the ergonomics of tip ejection have been re-imagined to relieve stress on the thumb and joints.
Halo™ Single Channel Pipettor, lightweight, 2-20µL
Caratteristiche e vantaggi
- 10-40% lighter than leading competitors
- 4 Digit micrometer with DriftGuard protection
- Exclusive 1250µL model for extended length tips
- User-enabled calibration port
- Compatible with universal tips
Note legali
Halo is a trademark of MTC Bio
MTC is a trademark of MTC Bio
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)