Key Documents
L6789
Laser Tough-Spots®
diam. 1 in., white
Sinonimo/i:
solvent resistant labels, temperature resistant labels
About This Item
Codice UNSPSC:
55121611
NACRES:
NB.35
Materiali
polyolefin
white
Confezionamento
pack of 1260 ea (63 labels per sheet)
Produttore/marchio commerciale
Diversified Biotech SPOT-4000
Diametro
1 in.
Categorie correlate
Descrizione generale
Tough-Spots are chemically inert, polyvinyl labels that adhere to all plastics. Tough-Spots withstand boiling water baths, conventional and cryogenic freezer temperatures, most organic solvents, caustic agents and other challenges without peeling, becoming brittle or degrading. Labels can be printed directly from the laser printer. 20 sheets per pack.
Note legali
Tough-Spots is a registered trademark of Diversified Biotech