HS120758
Box Top Dispenser for Parafilm
natural (dispenser)
About This Item
Prodotti consigliati
Materiali
acrylonitrile-butadiene-styrene (ABS)
natural (dispenser)
Caratteristiche
to hold 2 x 50 mm or 1 x 100 mm Parafilm M Sealing Film roll
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Heathrow Scientific 120758
Lungh. × Largh. × Alt.
123 mm × 172 mm × 99 mm
Descrizione generale
Caratteristiche e vantaggi
- Accommodates two 50 mm (2-inch), or one 100 mm (4-inch) Parafilm® M rolls
- Easy application, simply place the dispenser on top of the Parafilm® M box and pull the sealing film through, there is no need to remove rolls from the packaging
- Built in safety blade slides and cuts with no danger to fingertips and eliminates the need for scissors
- Supplied with two safety blades, additional replacement blades are available for purchase
- Made from ABS plastic making the unit easy to clean
Note legali
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
HS120758-1EA | 4065265819909 |