GBL622304
Grace Bio-Labs CoverWell™ perfusion chambers
4 chambers, port diam. 1.5 mm, chamber W × L × thickness 6.35 mm × 19 mm × 2.5 mm
Sinonimo/i:
leak proof enclosure for slides, microscope stick on multiwell chamber, reusable press to seal multiwell chamber, water tight multiple well chamber
About This Item
Prodotti consigliati
Materiali
clear polycarbonate backing
red silicon wells
Sterilità
non-sterile
Caratteristiche
wells 4
Confezionamento
pack of 20 ea
Produttore/marchio commerciale
Grace Bio-Labs 622304
Camera: largh. × lungh. × spessore
6.35 mm × 19 mm × 2.5 mm
Diametro porta
1.5 mm
Pozzetti
4
Categorie correlate
Descrizione generale
CoverWell perfusion chambers form removable, water-tight multiwell enclosures by pressing to coverglass or microscope slides. Chambers are made of medical-grade silicone bonded to clear, UV-transparent plastic, and are removable for permanent mounting. The thin polycarbonate cover displays excellent transparency. The perfusion chambers are ideal for live cell imaging on upright or inverted microscopes at high magnification.
Note legali
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
GBL622304-20EA | 4061832399508 |