Documenti fondamentali
GBL204908
Grace Bio-Labs FlexWell removable incubation chambers
L × W × thickness 6.5 mm × 6.5 mm × 3.2 mm, clear silicone, adhesive one side, wells, 8
Sinonimo/i:
flexible silicone slide gasket, silicone multiwell gasket
About This Item
Codice UNSPSC:
41122600
NACRES:
NB.22
Materiali
clear silicone
Sterilità
non-sterile
Caratteristiche
wells 8
Confezionamento
pack of 10 ea
Produttore/marchio commerciale
Grace Bio-Labs 204908
Lungh. × largh. × spess.
6.5 mm × 6.5 mm × 3.2 mm
Descrizione generale
FlexWell incubation chambers provide a convenient way to conduct binding reactions on protein microarrays using nitrocellulose coated ONCYTE® Film slides.
  • Silicone FlexWell forms wells on slides using a clean release adhesive to isolate up to 16 specimens per slide
  • Slides and FlexWell fit into a reusable slide tray organizing wells into the footprint of a SMS compliant microtiter plate
  • Gaskets may be sealed for mixing and to prevent evaporation
Note legali
FlexWell is a trademark of Grace Bio-Labs, Inc.
Oncyte is a registered trademark of Grace Bio-Labs, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)