EPM1001-0240
Platform for Eppendorf® New Brunswick™ Innova® 2000 Platform Shaker
universal, interchangeable, stainless steel
About This Item
Codice UNSPSC:
41103816
NACRES:
NB.73
Prodotti consigliati
Materiali
stainless steel
Caratteristiche
interchangeable
universal
Produttore/marchio commerciale
Eppendorf® M1001-0240 (Global)
Lungh. × largh.
33 cm × 28 cm
Categorie correlate
Descrizione generale
Platforms dedicated to fit Eppendorf New Brunswick Innova 2000 Platform Shakers
Note legali
Eppendorf is a registered trademark of Eppendorf AG
Innova is a registered trademark of Eppendorf, Inc.
New Brunswick is a trademark of Eppendorf AG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)