Documenti fondamentali
EP5391000001
Eppendorf(R) HeatSealer
S100
About This Item
Codice UNSPSC:
41102400
NACRES:
NC.21
Ingresso CA/CC
100 - 230 V, 50/60 Hz, EuroPlug
Caratteristiche
CE compliant
CSA approved
fixed Sealing, temperature
seals 96 and 384 well plates
Produttore/marchio commerciale
Eppendorf® 5391000001 (EU)
Descrizione generale
The Eppendorf HeatSealer Family offers you reliable and effective Sealing, of a variety of plates. You can choose from a more compact (S100) and a more flexible (S200) instrument according to your Sealing, needs.
Caratteristiche e vantaggi
Hermetic Sealing, of multiwell plates
Eliminates evaporation in PCR
Seals 96- and 384-well plates
Integrated thermostat prevents overheating
Improved mechanics for even easier Sealing,
PCR Plate Adapter included
Note legali
Eppendorf is a registered trademark of Eppendorf AG
Accessorio
N° Catalogo
Descrizione
Determinazione del prezzo
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)