Documenti fondamentali
EP1330
Sigma-Aldrich
Sigma-Aldrich® CVS10 replacement platinum wire
L × diam. 50 cm × 0.2 mm
About This Item
Codice UNSPSC:
41100000
NACRES:
NB.22
L × diam.
50 cm × 0.2 mm
Note legali
SIGMA-ALDRICH is a registered trademark of Merck KGaA, Darmstadt, Germany
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 5
1 of 5
Platinum wire, diam. 1.5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
349399
Platinum
Platinum wire reel, 0.05m, diameter 1.0mm, annealed, may be used as one component of a thermocouple, 99.99%
GF43288487
Platinum
Platinum insulated wire, 1m, conductor diameter 0.125mm, insulation thickness 0.016mm, PTFE (polytetrafluoroethylene) insulation, 99.99%
GF47215313
Platinum
Platinum wire, diam. 0.10 mm, 99.99% trace metals basis
Sigma-Aldrich
357367
Platinum
Platinum wire, diam. 0.127 mm, 99.99% trace metals basis
Sigma-Aldrich
267163
Platinum